Skip to Content
Merck
All Photos(2)

Key Documents

253049

Sigma-Aldrich

Potash, sulfurated

Synonym(s):

Dipotassium thiosulfate mixture with potassium sulfide, Liver of sulfur, Sulfurated potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4/(K2Sx)2
CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

form

chunks
lumps

Quality Level

composition

sulfide, ≥12.8%

SMILES string

[K+].[K+].[K]S[K].[O-]S([O-])(=O)=S

InChI

1S/4K.H2O3S2.S/c;;;;1-5(2,3)4;/h;;;;(H2,1,2,3,4);/q;;2*+1;;/p-2

InChI key

PFSBHPPVZHPSCB-UHFFFAOYSA-L

Application

Sulfurated potash may be used in the preparation of N-substituted thionamides such as (+)-(S)-thion-α-methylbutyramide, (+)-(S)-(thionhydratropyl)morpholine and (+)-(S)-N-methylthion-α-methylbutyramide.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical Rotatory Dispersion Studies. XCVII. Anomalous Rotatory Dispersion and Circular Dichroism Curves Associated with Thionamides. Application to Stereochemical Studies of Carboxylic Acids2
Burakevich JV & Djerassi C
Journal of the American Chemical Society, 87(1), 51-60 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service