Skip to Content
Merck
All Photos(1)

Key Documents

16704

Supelco

Butyrophenone

analytical standard

Synonym(s):

1-Benzoylpropane, 1-Phenyl-1-butanone, NSC 8463, Phenyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2CH2CH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
508305
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.519-1.521
n20/D 1.520 (lit.)

bp

228-230 °C (lit.)

mp

11-13 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCC(=O)c1ccccc1

InChI

1S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8H,2,6H2,1H3

InChI key

FFSAXUULYPJSKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service