Skip to Content
Merck
All Photos(1)

Key Documents

02369

Supelco

Furfuryl acetate

analytical reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical reference material
analytical standard

Assay

≥97.5% (qNMR)
≥98.5% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

impurities

≤0.2% water

refractive index

n20/D 1.460-1.464
n20/D 1.462 (lit.)

bp

175-177 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

SMILES string

CC(=O)OCc1ccco1

InChI

1S/C7H8O3/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

CKOYRRWBOKMNRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service