Skip to Content
Merck
All Photos(1)

Key Documents

27-0220

Sigma-Aldrich

Resorcinol

SAJ first grade, ≥98.0%

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

Assay

≥98.0%

form

solid

autoignition temp.

1126 °F

availability

available only in Japan

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphate-functionalized carbon monoliths from deep eutectic solvents and their use as monolithic electrodes in supercapacitors.
Daniel Carriazo et al.
ChemSusChem, 5(8), 1405-1409 (2012-07-06)
Antonio B Fuertes et al.
Chemical communications (Cambridge, England), 48(49), 6124-6126 (2012-05-15)
Core@shell spheres made up of a thin layer of resorcinol-formaldehyde enveloping a silica core were prepared by means of a one-step method under Stöber conditions. These spheres are used as a platform for the synthesis of carbon or polymeric capsules
Maurizio Scaltriti et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(17), 4508-4513 (2012-06-22)
Many kinases and hormone receptors, important for cancer cell proliferation and survival, bind to and are dependent on the Hsp90 cycle for their folding and maturation. This provides the rationale for the development of small-molecule ATP competitors that, inhibiting Hsp90
Fotini Karavassili et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 12), 1632-1641 (2012-11-16)
The effects of the ligands phenol and resorcinol on the crystallization of human insulin have been investigated as a function of pH. Powder diffraction data were used to characterize several distinct polymorphic forms. A previously unknown polymorph with monoclinic symmetry
Anton V Gulevich et al.
Journal of the American Chemical Society, 134(12), 5528-5531 (2012-03-15)
The efficient Pd-catalyzed double-fold C-H oxygenation of arenes into resorcinols using the newly developed 2-pyrimidyldiisopropylsilyl (PyrDipSi) directing group is described. Its use allows for the sequential introduction of OAc and OPiv groups in a one-pot manner to produce orthogonally protected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service