19-0970
Manganese(II) nitrate hexahydrate
JIS special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Mn(NO3)2 · 6H2O
Molecular Weight:
287.04
UNSPSC Code:
12352302
Assay:
≥98.0%
grade:
JIS special grade
form:
solid
Recommended Products
grade
JIS special grade
Assay
≥98.0%
form
solid
availability
available only in Japan
storage temp.
2-8°C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1C - STOT RE 2 Inhalation
Target Organs
Brain
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reza Mohassel et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 204, 232-240 (2018-06-25)
GCMO NSs were synthesized by the reaction of metal nitrate salts in the presence of stabilizing agent and PG, by a Pechini method. Citric acid, maleic acid, succinic acid and 1,3,5-benzenetricarboxylic acid were used as stabilizing agents. The structure, morphology
Lu Zhao et al.
Nature communications, 10(1), 1278-1278 (2019-03-22)
Although single-atomically dispersed metal-Nx on carbon support (M-NC) has great potential in heterogeneous catalysis, the scalable synthesis of such single-atom catalysts (SACs) with high-loading metal-Nx is greatly challenging since the loading and single-atomic dispersion have to be balanced at high
Dong Wook Kwon et al.
Environmental technology, 38(22), 2785-2792 (2017-01-04)
The effects of physicochemical properties of Mn-Ti catalysts on O
Syam Kandula et al.
Small (Weinheim an der Bergstrasse, Germany), 14(23), e1800291-e1800291 (2018-05-11)
Supercapacitors suffer from lack of energy density and impulse the energy density limit, so a new class of hybrid electrode materials with promising architectures is strongly desirable. Here, the rational design of a 3D hierarchical sandwich Co9 S8 /α-MnS@N-C@MoS2 nanowire
Biswa Nath Bhadra et al.
Nanoscale, 10(31), 15035-15047 (2018-07-28)
A series of metal-organic framework-74s (MOF-74s), composed of two different metallic species (Zn/Ni or Zn/Mn in various compositions), were synthesized, and Ni or MnO-doped carbonaceous materials were first prepared by pyrolysis of the MOFs under an inert environment for catalytic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service