Skip to Content
Merck
All Photos(1)

Key Documents

05-0810

Sigma-Aldrich

Calcium L-lactate pentahydrate

SAJ first grade, ≥97.0%

Synonym(s):

L-Lactic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH(OH)COO]2Ca · 5H2O
CAS Number:
Molecular Weight:
308.29
Beilstein:
4923285
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

SAJ first grade

Assay

≥97.0%

form

solid

availability

available only in Japan

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O

InChI

1S/2C3H6O3.Ca.5H2O/c2*1-2(4)3(5)6;;;;;;/h2*2,4H,1H3,(H,5,6);;5*1H2/q;;+2;;;;;/p-2/t2*2-;;;;;;/m00....../s1

InChI key

HPVJXNNKHRNBOY-VFGZHDCSSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ehsan Rezabeigi et al.
Materials science & engineering. C, Materials for biological applications, 40, 248-252 (2014-05-27)
More than four decades after the discovery of 45S5 Bioglass® as the first bioactive material, this composition is still one of the most promising materials in the tissue engineering field. Sol-gel-derived bioactive glasses generally possess improved properties over other bioactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service