Skip to Content
Merck
All Photos(2)

Key Documents

MABS1271

Sigma-Aldrich

Anti-EPCR Antibody, clone JRK1535

clone JRK1535, from mouse

Synonym(s):

Endothelial protein C receptor, Activated protein C receptor, APC receptor, CD201, Endothelial cell protein C receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

JRK1535, monoclonal

species reactivity

human

should not react with

mouse

technique(s)

flow cytometry: suitable
neutralization: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PROCR(10544)

Related Categories

General description

Endothelial protein C receptor (UniProt Q9UNN8; also known as Activated protein C receptor, APC receptor, CD201, Endothelial cell protein C receptor) is encoded by the PROCR (also known as EPCR) gene (Gene ID 10544) in human. The coagulation cascade is an evolutionary conserved pathway in vertebrates that maintains vascular integrity. The procoagulant activity of the tssue factor (TF) pathway is counterbalanced by the protein C (PC) anticoagulant pathway to avoid intravascular thrombosis. EPCR is a CD1d–like immune receptor that binds the γ-carboxyl glutamic acid-rich (Gla) domain of PC and markedly improves PC activation in response to thrombin binding to endothelial cell-expressed thrombomodulin. EPCR also serves as the co-receptor for activated PC (aPC) in vascular protective signaling mediated by activation of PAR1. In addition, EPCR is found on hematopoietic, neuronal and epithelial progenitor populations, as well as among highly aggressive basal-like breast cancer subtypes. Endothelial overexpression of EPCR attenuates metastasis, presumably by dampening thrombin generation that supports metastatic tumor cell survival. EPCR-dependent PAR1 activation by aPC also stimulates breast cancer cell migration and prevents apoptosis of lung cancer cells. EPCR-selected populations of breast cancer cells grow as non-adherent spheroids and have high tumorigenicity when injected at low cell numbers. EPCR is initially produced with a signal peptide (a.a. 1-17), the removal of which yields the mature protein with a large extracellular region (a.a. 18-210), followed by a transmembrane domain (a.a. 211-231) and a short cytoplasmic tail (a.a. 232-238).

Specificity

Clone JRK1535 blocked activated protein C (APC) from binding cell surface EPCR by targeting a conformational epitope at the extracellular domain (Liaw, P.C., et al. (2001). J. Biol. Chem. 276(11):8364-8370).

Immunogen

Epitope: Extracellular domain.
Recombinant human EPCR extracellular domain.

Application

Anti-EPCR Antibody, clone JRK1535 is an antibody against EPCR for use in Flow Cytometry, Neutralizing.
Flow Cytometry Analysis: 0.1 µg of this antibody from a representative lot detected EPCR in one million HeLa cells.
Flow Cytometry Analysis: A representative lot, when conjugated with fluorescein, detected Doxorubicin (Cat. No. 324380) treatment-induced downregulation of EPCR expression on the surface of HUVECs. Glutathione pretreatment prevented Doxorubicin-induced EPCR downregulation (Woodley-Cook, J., et al. (2006). Mol. Cancer Ther. 5(12):3303-3311).
Flow Cytometry Analysis: A representative lot blocked fluorescein-labeled APC from binding exogenously expressed human EPCR on the surface of HEK293 transfectants. When labeled with fluorescein, clone JRK1535 stained HEK293 transfectants expressing human EPCR, but not transfectants expressing murine EPCR (Liaw, P.C., et al. (2001). J. Biol. Chem. 276(11):8364-8370).
Neutralizing Analysis: A representative lot inhibited APC generation from acrolein-treated HUVECs exposed to defibrinated, recalcified plasma (Swystun, L.L., et al. (2011). J Thromb Haemost. 9(4):767-775).
Neutralizing Analysis: A representative lot selectively affected the survival of sorted EPCR-positive, but not EPCR-negative, MDA-MB-231 mammary fat pad (mfp) cells in cultures. Pretreatment of EPCR-positive cells with clone JRK1535 prior to xenografting also suppressed the tumor-initiating capacity of the EPCR-positive cells in mice vivo (Schaffner, F., et al. (2013). PLoS One. 8(4):e61071).
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Evaluated by Flow Cytometry in A549 cells.

Flow Cytometry Analysis: 0.1 µg of this antibody detected EPCR in one million A549 cells.

Target description

24.80 kDa calculated. Greater than 40 kDa observed due to glycosylation (Woodley-Cook, J., et al. (2006). Mol. Cancer Ther. 5(12):3303-3311).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Cai et al.
Thrombosis and haemostasis, 121(11), 1448-1463 (2021-03-12)
Activated protein C (APC) is an anticoagulant plasma serine protease which exhibits potent cytoprotective and anti-inflammatory activities. Here, we studied protective effects of APC on the proinflammatory function of polyinosinic:polycytidylic acid [poly(I:C)], a synthetic analog of viral double-stranded RNA, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service