Skip to Content
Merck
All Photos(1)

Key Documents

MABN1834

Sigma-Aldrich

Anti-phospho-Filamin-A (Ser2152) Antibody, clone PS2

clone PS2, from mouse

Synonym(s):

Filamin-A, FLN-A, Actin-binding protein 280, ABP-280, Alpha-filamin, Endothelial actin-binding protein, Filamin-1, Non-muscle filamin, phospho-Filamin-A (Ser2152)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PS2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer2152)

Gene Information

human ... FLNA (2316)

General description

Filamin-A (UniProt P21333; also known as ABP-280, Actin binding protein 280, Alpha-filamin, Endothelial actin-binding protein, Filamin-1, FLN-A, Non-muscle filamin) is encoded by the FLNA (also known as ABP, CSBS, CVD1, FLN, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD) gene (Gene ID 2316) in human. Filamin-A (FLN-A) is an actin-binding protein essential for cytoskeletal rearrangement and cell locomotion. Serine 2152 of FLN-A is a PKA phosphorylation target. FLN-A S2152 phosphorylation is essential for FLN-A-mediated recruitment of signaling proteins, adhesion molecules, and the vesicle-trafficking complex to the peripheral actin rim, so as to stabilize the cortical structure and epithelial membrane. Mutations in FLNA and ARFGEF2 genes cause periventricular heterotopia (PVH or PH), a congenital malformation of cortical development characterized by a reduced brain size and ectopic neuronal nodules along the ventricular neuroepithelium as a result of impaired initiation of neuronal migration. Suppression of either Arfgef2 or Mekk4 results in increased cellular FLN-A level and Ser2152 phosphorylation. In addition, Arfgef2 or Mekk4 deletion causes PVH in knockout mice.

Specificity

Ser2152 of human and murine Filamin-A isoform 1 (P21333-1 & Q8BTM8-1) corresponds to Ser2144 of human Filamin-A isoform 2 (P21333-2). Equivalent site is not present in murine spliced isoform 2.

Immunogen

Epitope: Filamin repeat 20
KLH-conjugated linear peptide corresponding to the Filamin repeat 20 of human phospho-Filamin-A (Ser2152).

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-phospho-Filamin-A (Ser2152) Antibody, clone PS2 is validated for use in Western Blotting, Immunocytochemistry and Immunofluorescence for the detection of phospho-Filamin-A (Ser2152).
Western Blotting Analysis: A representative lot detected enhanced FLN-A Ser2152 phosphorylation in multiple tissue extracts (liver, heart, brain, and skin) from new born (P0) and E16.5 embryos of Arfgef2/Big2 knockout mice when compared with stage-matched tissue samples from wild-type mice (Zhang, J., et al. (2012). J. Neurosci. 32(36):12619-12629).
Western Blotting Analysis: A representative lot detected cellular FLN-A pSer2152 phosphorylation induction upon serum, forskolin, or EphB2-Fc addition to serum-starved CHP100 human neuroblastoma cells (Zhang, J. et al. (2013). J. Neurosci. 33(40):15735-15746).
Immunocytochemistry Analysis: A representative lot detected cellular FLN-A pSer2152 phosphorylation induction in neuronal progenitors from frontal cortex of E13.5–E14.5 mouse embryos, CHP100 human neuroblastoma cells, and A7, but not FLN-A-deficient M2, human melanoma cells upon forskolin addition (Zhang, J. et al. (2013). J. Neurosci. 33(40):15735-15746).
Immunofluorescence Analysis: A representative lot detected FLN-A pSer2152 immunoreactivity extended to the proliferative zone (PZ) surface at the periventricular heterotopia (PVH) congenital malformation site in the forebrain (FB) of E17.5 MEKK-/- mouse embryos in addition to the FLN-A pSer2152 immunoreactivity in the intermediate zone (IZ) and cortical plate (CP) seen in both wild-type and MEKK-knockout embryos by fluorescent immunohistochemistry (Sarkisian, M.R., et al. (2006). Neuron. 52(5):789-801).

Quality

Evaluated by Western Blotting in mouse embryonic fibroblast lysate.

Western Blotting Analysis: A 1:125 dilution of this antibody detected Calyculin A-stimulated Filamin-A Ser2152 phosphorylation in mouse embryonic fibroblasts (MEFs).

Target description

~280 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service