Skip to Content
Merck
All Photos(2)

Key Documents

MABN1191

Sigma-Aldrich

Anti-Parvalbumin Antibody, clone 4E9.2

clone 4E9.2, from mouse

Synonym(s):

Parvalbumin alpha, Parvalbumin, PV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4E9.2, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PVALB(5816)
mouse ... Pvalb(19293)
rat ... Pvalb(25269)

General description

Parvalbumin alpha (UniProt P02625; also known as PV) is encoded by the Pvalb (also known as Pva) gene (Gene ID 25269) in rat species. Parvalbumin alpha is a small (a.a. 2-110; Met1 removed posttranslationally) calcium-binding protein. Parvalbumin alpha is composed almost entirely of two EF-hand domains (a.a. 39-74 & 78-110) and is structurally related to calmodulin and troponin C. Parvalbumin (PV) is present in GABAergic interneurons in the nervous system, especially the reticular thalamus. PV interneurons are a subset of inhibitory GABAergic neurons that control the excitability of post-synaptic pyramidal neurons. Dysfunction or loss of PV interneurons is implicated in cognitive deficits associated with a number of well known psychiatric disorders, including schizophrenia, Alzheimer’s disease, and depression.

Specificity

Clone 4E9.2 detects an epitope conserved among human, mouse, and rat species.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence within the N-terminal half of rat parvalbumin.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Parvalbumin in human kidney, rat kidney, and mouse cerebral cortex tissue sections.
This Anti-Parvalbumin Antibody, clone 4E9.2 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Parvalbumin.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Parvalbumin in 10 µg of mouse brain tissue lysate.

Target description

~12 kDa observed. 11.93 kDa (human), 11.80 kDa (mouse), 11.79 kDa (rat) calculated (Met1 removed). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahithi Attaluri et al.
Frontiers in aging neuroscience, 15, 1200445-1200445 (2023-07-10)
Extracellular vesicles (EVs) released by human-induced pluripotent stem cell (hiPSC)-derived neural stem cells (NSCs) have robust antiinflammatory and neurogenic properties due to therapeutic miRNAs and proteins in their cargo. Hence, hiPSC-NSC-EVs are potentially an excellent biologic for treating neurodegenerative disorders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service