Skip to Content
Merck
All Photos(2)

Key Documents

MABC535

Sigma-Aldrich

Anti-BIRC2/IAP-2, clone 1E1-1-10 Antibody

clone 1E1-1-10, from rat

Synonym(s):

Baculoviral IAP repeat-containing protein 2, Inhibitor of apoptosis protein 2, IAP-2, mIAP-2, mIAP2, cIAP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E1-1-10, monoclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BIRC2(329)

General description

BRC2/IAP-2, also known as IAP homolog B, Inhibitor of apoptosis protein 2 and Baculoviral IAP repeat-containing protein 2, and encoded by the gene BIRC2/API1/IAP2/MIHB/ RNF48, is a multi-functional protein which regulates not only caspases and apoptosis, but also modulates inflammatory signaling and immunity, mitogenic kinase signaling, and cell proliferation, as well as cell invasion and metastasis. BIRC2/IAP-2 also acts as an E3 ubiquitin-protein ligase regulating NF-kappa-B signaling and regulates both canonical and non-canonical pathways. Additionally, BIRC2/IAP-2 acts as an important regulator of innate immune signaling, and finally, BIRC2/IAP-2 plays an important role in the modulation of the cell cycle. BIRC2/IAP-21 is localized to nucleus except during apoptotic events. BIRC2/IAP-2 is expressed in fetal and adult tissues with a strong emphasis found in adult skeletal muscle, thymus, testis, ovary and pancreas with low or absent expression in blood and brain.

Immunogen

Epitope: This antibody recognizes the BIR 2 repeat region of BIRC2/IAP-2.
Linear peptide corresponding to the BIR 2 repeat region of mouse BIRC2/IAP-2.

Application

Anti-BIRC2/IAP-2 antibody, clone 1E1-1-10 is an antibody against BIRC2/IAP-2 for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 2 µg/mL from a representative lot detected BIRC2/IAP-2 in C2C12 cell lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected BIRC2/IAP-2 in HeLa and Raji cell lysate (Gerlach, B., et al. (2011). Nature. 471(7340):591-596.).

Western Blotting Analysis: A representative lot from an independent laboratory detected BIRC2/IAP-2 in MEF cell lysate (Gerlach, B., et al. (2011). Nature. 471(7340):591-596.).

Western Blotting Analysis: A representative lot from an independent laboratory detected BIRC2/IAP-2 in stable cell lines expressing EYFP cIAP1 RING, or EYFP cIAP1 RING (Silke, J., et al. (2005). Proc Natl Acad Sci USA. 102(45):16182-16187.).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected BIRC2/IAP-2 in 10 µg of NIH/3T3 cell lysate.

Target description

~65 kDa observed. The calculated molecular weight of this protein is 70 kDa; however, this protein has been observed at lower molecular weights (Gerlach, B., et al. (2011). Nature. 471(7340):591-596.; Silke, J., et al. (2005). Proc Natl Acad Sci USA. 102(45):16182-16187.).

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service