Skip to Content
Merck
All Photos(1)

Key Documents

821005

Sigma-Aldrich

Probumin® Bovine Serum Albumin Life Science Grade

≥98%, USA origin, ≤50 μg/g IgG, cell culture| mammalian: suitable, enzyme immunoassay: suitable, Serology: suitable

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801

product name

Probumin® Bovine Serum Albumin Life Science Grade, pkg of 5 kg

biological source

bovine serum

Quality Level

Assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pkg of 5 kg

manufacturer/tradename

Probumin®

origin

USA origin

technique(s)

cell culture | mammalian: suitable using
enzyme immunoassay: suitable

dilution

(Serology)

impurities

≤50 μg/g IgG

loss

≤5%

pH

6.8-7.2

solubility

water: soluble 40 mg/mL

cation traces

heavy metals: ≤20 ppm

foreign activity

Tested for Mycoplasma
Tested for Protease

storage temp.

2-8°C

General description

A high purity, low endotoxin and low IgG BSA powder preparation with near native albumin characteristics. This BSA is first isolated by a proprietary heat-shock/ organic solvent process and further purified by non solvent based methodologies to reduce endotoxins to very low levels resulting in a highly monomeric bovine albumin that retains many of the binding, transfer, and physical properties of native albumin. Purity is = 98%; typical purity is 100%. This BSA may be used for molecular biology applications.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin® BSA grades are distinguished by differences in production. The use of terms such as ‘Diagnostic Grade′ or ‘Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin® BSA grades using the specific reagents, conditions and constraints of their assay.

For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service