Skip to Content
Merck
All Photos(1)

Key Documents

06-106

Sigma-Aldrich

Anti-IGFBP-1 Antibody

serum, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGFBP1(3484)

Specificity

IGFBP-1

Immunogen

IGFBP-1 purified from human amniotic fluid

Application

Detect IGFBP-1 using this Anti-IGFBP-1 Antibody validated for use in IC, IP & WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

routinely evaluated by immunoblot on IGFBP-1 (Catalog #12-129)

Target description

30kDa

Physical form

Antiserum
Lyophilized

Storage and Stability

Lyophilized: 2 years at 4°C Rehydrated: 6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radioimmunoassay of a 26,000-dalton plasma insulin-like growth factor-binding protein: control by nutritional variables.
Busby, W H, et al.
The Journal of clinical endocrinology and metabolism, 67, 1225-1230 (1988)
C-M Lai et al.
The British journal of ophthalmology, 89(7), 911-916 (2005-06-21)
To generate a mouse model for slow progressive retinal neovascularisation through vascular endothelial growth factor (VEGF) upregulation. Transgenic mice were generated via microinjection of a DNA construct containing the human VEGF165 (hVEGF) gene driven by a truncated mouse rhodopsin promoter.
R G Richards et al.
Biology of reproduction, 52(3), 609-615 (1995-03-01)
Term decidual tissue consists of terminally differentiated decidual cells, bone marrow-derived cells, and fibroblast cells. Since undifferentiated decidual cells are fibroblast-like cells of the endometrial stroma, the possibility exists that the fibroblast cells in term decidua are undifferentiated decidual cells.
W H Busby et al.
The Journal of biological chemistry, 263(28), 14203-14210 (1988-10-05)
Human amniotic fluid has been shown to contain a protein that binds insulin-like growth factor I and II (IGF-I and IGF-II). Partially purified preparations of this protein have been reported to inhibit the biologic actions of the IGFs. In these
Saffold virus type 3 (SAFV-3) persists in HeLa cells.
Himeda, T; Hosomi, T; Okuwa, T; Muraki, Y; Ohara, Y
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service