Skip to Content
Merck
All Photos(1)

Key Documents

L-030

Supelco

Lurasidone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36N4O2S · HCl
CAS Number:
Molecular Weight:
529.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.O=C1[C@@H]2[C@H]3CC[C@H](C3)[C@@H]2C(=O)N1C[C@@H]4CCCC[C@H]4CN5CCN(CC5)c6nsc7ccccc67

InChI

1S/C28H36N4O2S.ClH/c33-27-24-18-9-10-19(15-18)25(24)28(34)32(27)17-21-6-2-1-5-20(21)16-30-11-13-31(14-12-30)26-22-7-3-4-8-23(22)35-29-26;/h3-4,7-8,18-21,24-25H,1-2,5-6,9-17H2;1H/t18-,19+,20-,21-,24+,25-;/m0./s1

InChI key

NEKCRUIRPWNMLK-SCIYSFAVSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

A certified solution standard suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, pharmaceutical research, or urine drug testing. Marketed under the trade name Latuda®, lurasidone is a new atypical antipsychotic developed for the treatment of schizophrenia.

Application


  • LurasiDonein Bipolar Depression Research: A study explored the pharmacodynamic properties of lurasidone, hypothesizing its efficacy in acute bipolar depression. This research provides a deep dive into the mechanistic actions of lurasidone, enhancing understanding in neuropharmacological studies and aiding in the development of more effective treatments for bipolar disorder (Fountoulakis et al., 2024).

  • Quantification of LurasiDonein Clinical Samples: Development and validation of a liquid chromatography-tandem mass spectrometry method for quantifying lurasiDonein dried blood spot samples was reported. This method facilitates easier and less invasive monitoring of lurasiDonelevels in patients, crucial for effective pharmacological research and ensuring therapeutic efficacy in treatment regimes (Rajadhyaksha and Londhe, 2023).

  • Novel Methodologies in Clinical Trials: Research introduced a novel method for deriving adverse event prevalence in randomized controlled trials, which could potentially improve the understanding of the benefit-risk ratio of drugs including lurasidone. This approach is particularly relevant for drug labels and regulatory submissions, ensuring safer and more effective clinical outcomes (Piacentino et al., 2024).

  • Pharmacological Properties of Lurasidone: A study investigated how lurasiDoneblocks the voltage-gated potassium channels of coronary arterial smooth muscle cells, offering insights into its broader pharmacological impacts. This research is vital for assessing potential cardiovascular side effects and optimizing dosing strategies to mitigate risks in patients treated with lurasiDone(Zhuang et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Latuda is a registered trademark of Dainippon Sumitomo Pharma Co., Ltd.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service