All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
Beilstein:
174414
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
221-225 °C (lit.)
SMILES string
OC(=O)C1c2ccccc2Oc3ccccc13
InChI
1S/C14H10O3/c15-14(16)13-9-5-1-3-7-11(9)17-12-8-4-2-6-10(12)13/h1-8,13H,(H,15,16)
InChI key
VSBFNCXKYIEYIS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A new method for assaying propantheline and its degradation product, xanthene-9-carboxylic acid using high-performance liquid chromatography.
B G Charles et al.
Journal of pharmaceutical sciences, 72(1), 96-98 (1983-01-01)
A C Blackburn et al.
Acta crystallographica. Section C, Crystal structure communications, 52 ( Pt 6), 1486-1488 (1996-06-15)
In xanthene-9-carboxylic acid, C14H10O3, hydrogen bonding is of the cyclic dimer type but involves two crystallographically inequivalent molecules and does not occur about a center of symmetry. The carboxylic H atoms are ordered. The dihedral angle (fold angle) of the
Lovely Angel Panamparambil Antony et al.
Organic letters, 15(17), 4552-4555 (2013-08-21)
6-Hydroxy-3-oxo-3H-xanthene-9-carboxylic acid is introduced as the first transition-metal-free carbon monoxide releasing molecule activated by visible light (photoCORM). This water-soluble fluorescein analogue releases carbon monoxide in both water and methanol upon irradiation at 500 nm. When selectively irradiated in the presence
Valerio D'Amore et al.
Epilepsia, 56(7), 1141-1151 (2015-06-05)
Glutamate and γ-aminobutyric acid (GABA) are the key neurotransmitter systems in the cortical-thalamocortical network, involved in normal and pathologic oscillations such as spike-wave discharges (SWDs), which characterize different forms of absence epilepsy. Metabotropic glutamate (mGlu) and GABA receptors are widely
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service