Skip to Content
Merck
All Photos(1)

Key Documents

W318418

Sigma-Aldrich

2-Acetyl-1-methylpyrrole

≥98%, FG

Synonym(s):

1-(1-methylpyrrol-2-yl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
FEMA Number:
3184
EC Number:
Council of Europe no.:
11373
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.046
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.542 (lit.)

bp

200-202 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy

SMILES string

CC(=O)c1cccn1C

InChI

1S/C7H9NO/c1-6(9)7-4-3-5-8(7)2/h3-5H,1-2H3

InChI key

NZFLWVDXYUGFAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effect of the pyrrole polymerization mechanism on the antioxidative activity of nonenzymatic browning reactions.: Investigates how the polymerization of pyrrole compounds, including 2-Acetyl-1-methylpyrrole, influences their antioxidative properties in browning reactions. This has implications for food chemistry and the development of antioxidants (Hidalgo et al., 2003).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service