Skip to Content
Merck
All Photos(1)

Key Documents

91692

Sigma-Aldrich

2,2,2-Trifluoroethylamine

≥98.0% (GC)

Synonym(s):

2-Amino-1,1,1-trifluoroethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2NH2
CAS Number:
Molecular Weight:
99.06
Beilstein:
1733204
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

~7.6 psi ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.301 (lit.)

bp

36-37 °C (lit.)

density

1.262 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

NCC(F)(F)F

InChI

1S/C2H4F3N/c3-2(4,5)1-6/h1,6H2

InChI key

KIPSRYDSZQRPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Direct preparation of 2,2,2-trifluorodiazoethane useful for esterifying sulfonic acids

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.O. Meese
Synthesis, 1041-1041 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service