665185
Bis(chlorogold(I)) bis(diphenylphosphino)methane
97%
Synonym(s):
Methylenebis[diphenyl]phosphine gold complex
About This Item
Recommended Products
Assay
97%
form
solid
reaction suitability
core: gold
reagent type: catalyst
SMILES string
Cl[Au][PH](C[PH]([Au]Cl)(c1ccccc1)c2ccccc2)(c3ccccc3)c4ccccc4
InChI
1S/C25H22P2.2Au.2ClH/c1-5-13-22(14-6-1)26(23-15-7-2-8-16-23)21-27(24-17-9-3-10-18-24)25-19-11-4-12-20-25;;;;/h1-20H,21H2;;;2*1H/q;2*+1;;/p-2
InChI key
JNPUWTQJFLJEJC-UHFFFAOYSA-L
Application
- Pharmacological studies involving the anticancer activity of gold(I)-phosphine complexes as autophagy-inducing agents
- Triplet luminescent light emitting diodes with low turn-on voltage
Catalyst for:
- Stereoselective preparation of vinyltetrahydrofuran via intramolecular hydroalkoxylation
- Hydroamination and hydroalkoxylation reactions
- Asymmetric transition metal catalysis
- Gold(I) catalyzed intramolecular Schmidt reactions of homopropargyl azides
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
We are proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service