Skip to Content
Merck
All Photos(2)

Documents

458783

Sigma-Aldrich

1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea

95%

Synonym(s):

1,3-Bis(benzyloxycarbonyl)-2-methylisothiourea, 2-Methyl-1,3-di-Z-isothiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O2CN=C(SCH3)NHCO2CH2C6H5
CAS Number:
Molecular Weight:
358.41
Beilstein:
9211307
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

60-63 °C (lit.)

SMILES string

CS\C(NC(=O)OCc1ccccc1)=N/C(=O)OCc2ccccc2

InChI

1S/C18H18N2O4S/c1-25-16(19-17(21)23-12-14-8-4-2-5-9-14)20-18(22)24-13-15-10-6-3-7-11-15/h2-11H,12-13H2,1H3,(H,19,20,21,22)

InChI key

CGAMNSKIHXUDMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Shimokawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(23), 6878-6888 (2005-09-15)
Asymmetric total synthesis of batzelladine A (1) and batzelladine D (2) has been achieved. Our synthesis of batzelladines features 1) stereoselective construction of the cyclic guanidine system by means of successive 1,3-dipolar cycloaddition reaction and subsequent cyclization, 2) direct esterification
David M Raffel et al.
Journal of medicinal chemistry, 50(9), 2078-2088 (2007-04-11)
The norepinephrine transporter (NET) substrates [123I]-m-iodobenzylguanidine (MIBG) and [11C]-m-hydroxyephedrine (HED) are used as markers of cardiac sympathetic neurons and adrenergic tumors (pheochromocytoma, neuroblastoma). However, their rapid NET transport rates limit their ability to provide accurate measurements of cardiac nerve density.
Application of molecular topology to the search of novel NSAIDs: experimental validation of activity.
Galvez-Llompart M, et al.
Letters in Drug Design & Discovery, 7(6), 438-445 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service