HPLC Analysis of Amyloid β-Protein Fragment 10-35 on Discovery® BIO Wide Pore C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 25 cm × 4.6 mm I.D., 5 μm particles (568223-U)
mobile phase
[A] 0.1% TFA in water; [B] 0.1% TFA in water:0.1% TFA in acetonitrile; (25:75)
flow rate
1.3 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
10 μL, 1 mg/mL in water
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany