Merck
  • Structural and functional analysis of the GABARAP interaction motif (GIM).
Structural and functional analysis of the GABARAP interaction motif (GIM).
EMBO reports (2017-06-29)
Vladimir V Rogov, Alexandra Stolz, Arvind C Ravichandran, Diana O Rios-Szwed, Hironori Suzuki, Andreas Kniss, Frank Löhr, Soichi Wakatsuki, Volker Dötsch, Ivan Dikic, Renwick Cj Dobson, David G McEwan
ABSTRACT
Through the canonical LC3 interaction motif (LIR), [W/F/Y]-X1-X2-[I/L/V], protein complexes are recruited to autophagosomes to perform their functions as either autophagy adaptors or receptors. How these adaptors/receptors selectively interact with either LC3 or GABARAP families remains unclear. Herein, we determine the range of selectivity of 30 known core LIR motifs towards individual LC3s and GABARAPs. From these, we define a G ABARAP I nteraction M otif (GIM) sequence ([W/F]-[V/I]-X2-V) that the adaptor protein PLEKHM1 tightly conforms to. Using biophysical and structural approaches, we show that the PLEKHM1-LIR is indeed 11-fold more specific for GABARAP than LC3B. Selective mutation of the X1 and X2 positions either completely abolished the interaction with all LC3 and GABARAPs or increased PLEKHM1-GIM selectivity 20-fold towards LC3B. Finally, we show that conversion of p62/SQSTM1, FUNDC1 and FIP200 LIRs into our newly defined GIM, by introducing two valine residues, enhances their interaction with endogenous GABARAP over LC3B. The identification of a GABARAP-specific interaction motif will aid the identification and characterization of the expanding array of autophagy receptor and adaptor proteins and their in vivo functions.
Sigma-Aldrich
Anti-PLEKHM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Roche
Protein A Agarose, >98% (HPLC and SDS-PAGE), suspension