Merck
Characterization of Reproductive Dormancy in Male
Frontiers in physiology (2016-12-10)
Olga I Kubrak, Lucie Kučerová, Ulrich Theopold, Sören Nylin, Dick R Nässel
ABSTRACT
Insects are known to respond to seasonal and adverse environmental changes by entering dormancy, also known as diapause. In some insect species, including
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol