Merck
  • A new candidate substrate for cell-matrix adhesion study: the acellular human amniotic matrix.
A new candidate substrate for cell-matrix adhesion study: the acellular human amniotic matrix.
Journal of biomedicine & biotechnology (2012-10-24)
Qianchen Guo, Xuya Lu, Yuan Xue, Hong Zheng, Xiaotao Zhao, Huajian Zhao
ABSTRACT
In vivo adhesions between cells and the extracellular matrix play a crucial role in cell differentiation, proliferation, and migration as well as tissue remodeling. Natural three-dimensional (3D) matrices, such as self-assembling matrices and Matrigel, have limitations in terms of their biomechanical properties. Here, we present a simple method to produce an acellular human amniotic matrix (AHAM) with preserved biomechanical properties and a favorable adhesion potential. On the stromal side of the AHAM, human foreskin fibroblasts (HFFs) attached and extended with bipolar spindle-shaped morphology proliferated to multilayer networks, invaded into the AHAM, and migrated in a straight line. Moreover, αV integrin, paxillin, and fibronectin were observed to colocalize after 24 h of HFF culture on the stromal side of the AHAM. Our results indicate that the AHAM may be an ideal candidate as a cell-matrix adhesion substrate to study cell adhesion and invasion as well as other functions in vitro under a tensile force that mimics the in vivo environment.
Sigma-Aldrich
Anti-TGF β Receptor II antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Collagen II antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Collagen IV α2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Collagen III antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TGF β Receptor I antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone IST-4, ascites fluid
Sigma-Aldrich
Anti-Collagen I α2 antibody produced in rabbit, affinity isolated antibody