Merck
  • Proton-shuttling lichen compound usnic acid affects mitochondrial and lysosomal function in cancer cells.
Proton-shuttling lichen compound usnic acid affects mitochondrial and lysosomal function in cancer cells.
PloS one (2012-12-12)
Margret Bessadottir, Mar Egilsson, Eydis Einarsdottir, Iris H Magnusdottir, Margret H Ogmundsdottir, Sesselja Omarsdottir, Helga M Ogmundsdottir
ABSTRACT
The lichen compound usnic acid (UA) is a lipophilic weak acid that acts as a proton shuttle and causes loss of mitochondrial inner membrane potential. In the current study we show that UA treatment induced the formation of autophagosomes in human cancer cells, but had minimal effects on normal human fibroblasts. However, autophagic flux was incomplete, degradation of autophagosomal content did not occur and acidification was defective. UA-treated cells showed reduced ATP levels and activation of AMP kinase as well as signs of cellular stress. UA is thus likely to trigger autophagosome formation both by energy depletion and stress conditions. Our findings indicate that the H(+)-shuttling effect of UA operates not only in mitochondria as previously shown, but also in lysosomes, and have implications for therapeutic manipulation of autophagy and pH-determined drug distribution.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(+)-Usnic acid, 98%
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution