Merck
  • A Mutation in Syne2 Causes Early Retinal Defects in Photoreceptors, Secondary Neurons, and Müller Glia.
A Mutation in Syne2 Causes Early Retinal Defects in Photoreceptors, Secondary Neurons, and Müller Glia.
Investigative ophthalmology & visual science (2015-06-13)
Dennis M Maddox, Gayle B Collin, Akihiro Ikeda, C Herbert Pratt, Sakae Ikeda, Britt A Johnson, Ron E Hurd, Lindsay S Shopland, Jürgen K Naggert, Bo Chang, Mark P Krebs, Patsy M Nishina
ABSTRACT
The purpose of this study was to identify the molecular basis and characterize the pathological consequences of a spontaneous mutation named cone photoreceptor function loss 8 (cpfl8) in a mouse model with a significantly reduced cone electroretinography (ERG) response. The chromosomal position for the recessive cpfl8 mutation was determined by DNA pooling and by subsequent genotyping with simple sequence length polymorphic markers in an F2 intercross phenotyped by ERG. Genes within the candidate region of both mutants and controls were directly sequenced and compared. The effects of the mutation were examined in longitudinal studies by light microscopy, marker analysis, transmission electron microscopy, and ERG. The cpfl8 mutation was mapped to Chromosome 12, and a premature stop codon was identified in the spectrin repeat containing nuclear envelope 2 (Syne2) gene. The reduced cone ERG response was due to a significant reduction in cone photoreceptors. Longitudinal studies of the early postnatal retina indicated that the cone photoreceptors fail to develop properly, rod photoreceptors mislocalize to the inner nuclear layer, and both rods and cones undergo apoptosis prematurely. Moreover, we observed migration defects of secondary neurons and ectopic Müller cell bodies in the outer nuclear layer in early postnatal development. SYNE2 is important for normal retinal development. We have determined that not only is photoreceptor nuclear migration affected, but also the positions of Müller glia and secondary neurons are disturbed early in retinal development. The cpfl8 mouse model will serve as an important resource for further examining the role of nuclear scaffolding and migration in the developing retina.
Sigma-Aldrich
MISSION® esiRNA, targeting human SYNE2
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
SAFC
Tricine
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)