Merck
Control of I-V hysteresis in CH3NH3PbI3 perovskite solar cell.
The journal of physical chemistry letters (2015-11-10)
Hui-Seon Kim, In-Hyuk Jang, Namyoung Ahn, Mansoo Choi, Antonio Guerrero, Juan Bisquert, Nam-Gyu Park
ABSTRACT
Mismatch of current (I)-voltage (V) curves with respect to the scan direction, so-called I-V hysteresis, raises critical issue in MAPbI3 (MA = CH3NH3) perovskite solar cell. Although ferroelectric and ion migration have been proposed as a basis for the hysteresis, origin of hysteresis has not been apparently unraveled. We report here on the origin of I-V hysteresis of perovskite solar cell that was systematically evaluated by the interface-dependent electrode polarizations. Frequency (f)-dependent capacitance (C) revealed that the normal planar structure with the TiO2/MAPbI3/spiro-MeOTAD configuration showed most significant I-V hysteresis along with highest capacitance (10(-2) F/cm(2)) among the studied cell configurations. Substantial reduction in capacitance to 10(-3) F/cm(2) was observed upon replacing TiO2 with PCBM, indicative of the TiO2 layer being mainly responsible for the hysteresis. The capacitance was intensively reduced to 10(-5) F/cm(2) and C-f feature shifted to higher frequency for the hysteresis-free planar structures with combination of PSS, NiO, and PCBM, which underlines the spiro-MeOTAD in part contributes to the hysteresis. This work is expected to provide a key to the solution of the problem on I-V hysteresis in perovskite solar cell.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%