Merck
  • Physiological relevant in vitro evaluation of polymer coats for gastroretentive floating tablets.
Physiological relevant in vitro evaluation of polymer coats for gastroretentive floating tablets.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-08-03)
Friederike Eisenächer, Grzegorz Garbacz, Karsten Mäder
ABSTRACT
Gastroretentive drug delivery systems are retained in the stomach for a sufficient time interval, releasing the drug in a controlled manner. According to literature, the floating principle is the most frequently used formulation approach for gastric retention. However, many publications lack information of the floating forces, the impact of different pH-values and almost no information exist concerning the resistance of the floating performance against physiological relevant stress. Therefore, we evaluated the performance of CO2-generating floating bilayer (drug and floating layer) tablets with respect to robustness, drug release profile, pH dependence and floating behaviour. Bilayer tablets were coated with a flexible and water permeable, but CO2-retaining polymer film of either polyvinyl acetate or ammonio-methacrylate copolymer type A. Metformin-HCl was used as a relevant model drug due to its dose-dependent and saturable absorption from the proximal part of the small intestine. To mimic physiological relevant mechanical stress conditions, recently developed dissolution stress tests with pulsed pressures were applied in addition to release studies according to the pharmacopeia. Bilayer tablets coated with polyvinyl acetate showed short floating lag times, reasonable floating strength values, floating durations of more than 24h in simulated gastric fluid and a robust and pH independent release of Metformin-HCl. Tablets coated with ammonio-methacrylate copolymer type A showed a higher permeability for the active ingredient combined with a decreased robustness of the inflated tablets. Both polymers can be used for balloon-like floating devices. The appropriate polymer has to be chosen dependent from the properties of the active ingredient and requested application of the delivery device. Furthermore, the dissolution stress test analysis is able to indicate possible safety issues of gastroretentive formulations as well as to characterise the robustness of formulation principles towards mechanical stresses of bio-relevant intensity.
Sigma-Aldrich
Triacetin, 99%
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Triacetin, 99%, FCC, FG
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium stearate, technical grade
Supelco
Triacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triacetin, analytical standard
USP
Triacetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis