Merck
  • Comparison of the sensitivity and specificity of 5 image sets of dual-energy computed tomography for detecting first-pass myocardial perfusion defects compared with positron emission tomography.
Comparison of the sensitivity and specificity of 5 image sets of dual-energy computed tomography for detecting first-pass myocardial perfusion defects compared with positron emission tomography.
Medicine (2014-12-20)
Wenhuan Li, Xiaolian Zhu, Jing Li, Cheng Peng, Nan Chen, Zhigang Qi, Qi Yang, Yan Gao, Yang Zhao, Kai Sun, Kuncheng Li
ABSTRACT
The sensitivity and specificity of 5 different image sets of dual-energy computed tomography (DECT) for the detection of first-pass myocardial perfusion defects have not systematically been compared using positron emission tomography (PET) as a reference standard. Forty-nine consecutive patients, with known or strongly suspected of coronary artery disease, were prospectively enrolled in our study. Cardiac DECT was performed at rest state using a second-generation 128-slice dual-source CT. The DECT data were reconstructed to iodine maps, monoenergetic images, 100 kV images, nonlinearly blended images, and linearly blended images by different postprocessing techniques. The myocardial perfusion defects on DECT images were visually assessed by 5 observers, using standard 17-segment model. Diagnostic accuracy of 5 image sets was assessed using nitrogen-13 ammonia PET as the gold standard. Discrimination was quantified using the area under the receiver operating characteristic curve (AUC), and AUCs were compared using the method of DeLong. The DECT and PET examinations were successfully completed in 30 patients and a total of 90 territories and 510 segments were analyzed. Cardiac PET revealed myocardial perfusion defects in 56 territories (62%) and 209 segments (41%). The AUC of iodine maps, monoenergetic images, 100 kV images, nonlinearly blended images, and linearly blended images were 0.986, 0.934, 0.913, 0.881, and 0.871, respectively, on a per-territory basis. These values were 0.922, 0.813, 0.779, 0.763, and 0.728, respectively, on a per-segment basis. DECT iodine maps shows high sensitivity and specificity, and is superior to other DECT image sets for the detection of myocardial perfusion defects in the first-pass myocardial perfusion.
Product Description
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Metoprolol tartrate, United States Pharmacopeia (USP) Reference Standard
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder