Merck
  • New isoquinolinequinone alkaloids from the South China Sea nudibranch Jorunna funebris and its possible sponge-prey Xestospongia sp.
New isoquinolinequinone alkaloids from the South China Sea nudibranch Jorunna funebris and its possible sponge-prey Xestospongia sp.
Fitoterapia (2014-04-29)
Wen-Fei He, Yan Li, Mei-Tang Feng, Margherita Gavagnin, Ernesto Mollo, Shui-Chun Mao, Yue-Wei Guo
ABSTRACT
Two new renieramycin-type bistetrahydroisoquinolinequinone alkaloids, fennebricins A (1) and B (5), and one new isoquinolinequinone alkaloid, N-formyl-1,2-dihydrorenierol (7), were isolated from the skin of the South China Sea nudibranch Jorunna funebris and its possible sponge-prey Xestospongia sp., together with eight known metabolites, including three bistetrahydroisoquinolinequinones (2-4) and five isoquinolinequinones (8-12). Their structures were elucidated by analysis of spectroscopic data including 1D and 2D NMR and high-resolution electrospray ionization mass spectrometry (HRESIMS) and by comparison with data for related known compounds. All the metabolites except for 7 occurred simultaneously in the two animals, supporting recent ecological studies that the nudibranch J. funebris preys on the sponge of the genus Xestospongia.
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
p-Anisaldehyde, 98%
Supelco
p-Anisaldehyde, analytical standard
Sigma-Aldrich
p-Anisaldehyde, natural, FG