Merck
  • The Moraxella catarrhalis-induced pro-inflammatory immune response is enhanced by the activation of the epidermal growth factor receptor in human pulmonary epithelial cells.
The Moraxella catarrhalis-induced pro-inflammatory immune response is enhanced by the activation of the epidermal growth factor receptor in human pulmonary epithelial cells.
Biochemical and biophysical research communications (2014-07-01)
Philippe Dje N'Guessan, Helge Haarmann, Tamara Steiner, Kerstin Heyl, Frauke Schreiber, Annina Heinrich, Hortense Slevogt
ABSTRACT
Chronic lower airway inflammation is considered to be a major cause of pathogenesis and disease progression in chronic obstructive pulmonary disease (COPD). Moraxella catarrhalis is a COPD-associated pathogen causing exacerbations and bacterial colonization in the lower airways of patients, which may contribute to chronic inflammation. Increasing evidence suggests that the epidermal growth factor receptor (EGFR) modulates inflammatory processes in the human airways. The goal of this study was to investigate the role of EGFR in the M. catarrhalis-induced pro-inflammatory immune response in airway epithelial cells. The effects of inhibition and gene silencing of EGFR on M. catarrhalis-dependent pro-inflammatory cytokine expression in human primary bronchial epithelial cells (NHBEs), as well as the pulmonary epithelial cell lines BEAS-2B and A549 were analyzed. We also assessed the involvement of EGFR-dependent ERK and NF-κB signaling pathways. The M. catarrhalis-induced pro-inflammatory immune response depends, at least in part, on the phosphorylation and activation of the EGF receptor. Interaction of M. catarrhalis with EGFR increases the secretion of pro-inflammatory cytokines, which is mediated via ERK and NF-κB activation. The interaction between M. catarrhalis and EGFR increases airway inflammation caused by this pathogen. Our data suggest that the inhibition of EGFR signaling in COPD could be an interesting target for reducing M. catarrhalis-induced airway inflammation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Egfr
Sigma-Aldrich
MISSION® esiRNA, targeting human EGFR
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Tyrosine, 99%