Merck
  • Histone deacetylase inhibitor valproic acid promotes the differentiation of human induced pluripotent stem cells into hepatocyte-like cells.
Histone deacetylase inhibitor valproic acid promotes the differentiation of human induced pluripotent stem cells into hepatocyte-like cells.
PloS one (2014-08-02)
Yuki Kondo, Takahiro Iwao, Sachimi Yoshihashi, Kayo Mimori, Ruri Ogihara, Kiyoshi Nagata, Kouichi Kurose, Masayoshi Saito, Takuro Niwa, Takayoshi Suzuki, Naoki Miyata, Shigeru Ohmori, Katsunori Nakamura, Tamihide Matsunaga
ABSTRACT
In this study, we aimed to elucidate the effects and mechanism of action of valproic acid on hepatic differentiation from human induced pluripotent stem cell-derived hepatic progenitor cells. Human induced pluripotent stem cells were differentiated into endodermal cells in the presence of activin A and then into hepatic progenitor cells using dimethyl sulfoxide. Hepatic progenitor cells were matured in the presence of hepatocyte growth factor, oncostatin M, and dexamethasone with valproic acid that was added during the maturation process. After 25 days of differentiation, cells expressed hepatic marker genes and drug-metabolizing enzymes and exhibited drug-metabolizing enzyme activities. These expression levels and activities were increased by treatment with valproic acid, the timing and duration of which were important parameters to promote differentiation from human induced pluripotent stem cell-derived hepatic progenitor cells into hepatocytes. Valproic acid inhibited histone deacetylase activity during differentiation of human induced pluripotent stem cells, and other histone deacetylase inhibitors also enhanced differentiation into hepatocytes. In conclusion, histone deacetylase inhibitors such as valproic acid can be used to promote hepatic differentiation from human induced pluripotent stem cell-derived hepatic progenitor cells.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Ethosuximide
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Nifedipine, European Pharmacopoeia (EP) Reference Standard
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
USP
Ethosuximide, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Ethosuximide, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethosuximide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Caffeine, BioXtra