Merck
Synergistic in vitro antimalarial activity of omeprazole and quinine.
Antimicrobial agents and chemotherapy (1999-05-01)
T Skinner-Adams, T M Davis
ABSTRACT
Previous studies have shown that the proton pump inhibitor omeprazole has antimalarial activity in vitro. The interactions of omeprazole with commonly used antimalarial drugs were assessed in vitro. Omeprazole and quinine combinations were synergistic; however, chloroquine and omeprazole combinations were antagonistic. Artemisinin drugs had additive antimalarial activities with omeprazole.
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)