Merck
Paracetamol and phenacetin.
Drugs (1986-01-01)
S P Clissold
ABSTRACT
Since their synthesis in the late 1800s paracetamol (acetaminophen) and phenacetin have followed divergent pathways with regard to their popularity as mild analgesic/antipyretic drugs. Initially, paracetamol was discarded in favour of phenacetin because the latter drug was supposedly less toxic. Today the opposite is true, and paracetamol, along with aspirin, has become one of the two most popular 'over-the-counter' non-narcotic analgesic agents. This marked increase in the wide approval attained by paracetamol has been accompanied by the virtual commercial demise of phenacetin because of its role, albeit somewhat circumstantial, in causing analgesic nephropathy. Both paracetamol and phenacetin are effective mild analgesics, suitable for treating mild to moderate pain, and their actions are broadly comparable with those of aspirin and related salicylates, although they do not appear to possess significant anti-inflammatory activity. Since a major portion of a dose of phenacetin is rapidly metabolised to paracetamol, it seems possible that phenacetin owes some of its therapeutic activity to its main metabolite, paracetamol, whereas its most troublesome side effect (methaemoglobinaemia) is due to another metabolite, p-phenetidine. The mechanism of action of paracetamol is poorly defined, although it has been speculated that it may selectively inhibit prostaglandin production in the central nervous system, which would account for its analgesic/antipyretic properties. The lack of any significant influence on peripheral cyclooxygenase would explain the absence of anti-inflammatory activity. At therapeutic doses paracetamol is well tolerated and produces fewer side effects than aspirin. The most frequently reported adverse effect associated with paracetamol is hepatotoxicity, which occurs after acute overdosage (usually doses greater than 10 to 15g are needed) and, very rarely, during long term treatment with doses at the higher levels of the therapeutic range. Paracetamol damages the liver through the formation of a highly reactive metabolite which is normally inactivated by conjugation with glutathione. Overdoses of paracetamol exhaust glutathione stores, thus allowing the accumulation of this toxic metabolite which covalently binds with vital cell elements and can result in liver necrosis. Glutathione precursors (notably intravenous N-acetylcysteine) have proved remarkably successful in treating paracetamol overdose, as long as treatment is initiated within 10 hours.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material