Merck
Levetiracetam: a comprehensive review.
Expert review of neurotherapeutics (2010-02-09)
Amy Z Crepeau, David M Treiman
ABSTRACT
Levetiracetam was approved by the US FDA in 1999 after failing the traditional screening tests for antiepileptic drugs. In the 10 years since its introduction, it has become one of the first-line antiepileptic agents and has been evaluated in small uncontrolled studies and case series as an off-label treatment for status epilepticus, for nonepileptic neurological conditions, such as movement disorders, neuropathic pain and headaches, and for some psychiatric conditions. It has not been approved for any indication other than chronic epilepsy. A significant factor in using levetiracetam for seizures and other disorders has been its ease of dosing and tolerability. In this article, we outline how levetiracetam was discovered, the animal and human data showing its antiepileptic activity, its potential use in other neurological conditions and the significant adverse events. We also discuss the direction antiepileptic drug development will take in the future.
Product Description
Levetiracetam, European Pharmacopoeia (EP) Reference Standard
Supelco
Levetiracetam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Supelco
Levetiracetam, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Levetiracetam, United States Pharmacopeia (USP) Reference Standard