Merck
  • An updated unified pharmacophore model of the benzodiazepine binding site on gamma-aminobutyric acid(a) receptors: correlation with comparative models.
An updated unified pharmacophore model of the benzodiazepine binding site on gamma-aminobutyric acid(a) receptors: correlation with comparative models.
Current medicinal chemistry (2007-11-30)
T Clayton, J L Chen, M Ernst, L Richter, B A Cromer, C J Morton, H Ng, C C Kaczorowski, F J Helmstetter, R Furtmüller, G Ecker, M W Parker, W Sieghart, J M Cook
ABSTRACT
A successful unified pharmacophore/receptor model which has guided the synthesis of subtype selective compounds is reviewed in light of recent developments both in ligand synthesis and structural studies of the binding site itself. The evaluation of experimental data in combination with a comparative model of the alpha1beta2gamma2 GABA(A) receptor leads to an orientation of the pharmacophore model within the Bz BS. Results not only are important for the rational design of selective ligands, but also for the identification and evaluation of possible roles which specific residues may have within the benzodiazepine binding pocket.
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard