Merck
  • Amantadine neurotoxicity in a pediatric patient with renal insufficiency.
Amantadine neurotoxicity in a pediatric patient with renal insufficiency.
DICP : the annals of pharmacotherapy (1991-11-01)
D K Strong, D D Eisenstat, S M Bryson, D S Sitar, G S Arbus
ABSTRACT
Amantadine hydrochloride, a dopamine agonist with antiviral and antiparkinsonism properties, is used for the prevention and treatment of influenza A respiratory infections in high-risk populations. The occurrence of amantadine-induced hallucinations and tremors is described in a young, renal transplant patient with declining renal function. Following discontinuation of amantadine, plasma amantadine concentrations were correlated with central nervous system toxicity. In view of the usage of amantadine in renal transplant recipients and the elderly, clinicians must be alert to the possibility of amantadine-induced neurotoxicity in patients with changing renal function.
Product Description
Loratadine, European Pharmacopoeia (EP) Reference Standard
Loratadine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Loratadine, ≥98% (HPLC), powder