Merck
  • Failure of high-dose ergocalciferol to correct vitamin D deficiency in adults with cystic fibrosis.
Failure of high-dose ergocalciferol to correct vitamin D deficiency in adults with cystic fibrosis.
American journal of respiratory and critical care medicine (2005-04-30)
Michael P Boyle, Michelle L Noschese, Sharon L Watts, Marsha E Davis, Shane E Stenner, Noah Lechtzin
ABSTRACT
Treatment guidelines for vitamin D monitoring and supplementation in cystic fibrosis (CF) have recently been developed and published by a consensus committee, but have not been prospectively tested. To use these guidelines to determine the percentage of adults with CF requiring vitamin D repletion therapy and to evaluate the effectiveness of the currently recommended high-dose oral ergocalciferol repletion protocol. Prospective study of clinical outcomes after therapy with the recommended vitamin D repletion algorithm. Of 134 adults with CF, 109 (81.3%) were found to have 25-hydroxyvitamin D (25-OHD) levels below the recommended 30 ng/ml. Sixty-six of these adults completed the recommended course of 400,000 IU of oral ergocalciferol over 2 months, and only five (8%) responded with correction of their serum 25-OHD to the goal of 30 ng/ml or greater (mean change, +0.3 ng/ml; from 18.8 to 19.1 ng/ml). In the 33 adults with CF who also completed the recommended second course of 800,000 IU of ergocalciferol over 2 months, none demonstrated correction of their deficiency (mean change, -1.2 ng/ml). The results of this study demonstrate that a majority of adults with CF have serum 25-OHD levels below 30 ng/ml, and the currently recommended ergocalciferol repletion regimen often does not fully correct vitamin D deficiency and may need to be revised to include even higher dosing of ergocalciferol. Further work is needed to establish the ideal 25-OHD level for maximizing calcium absorption and bone health in CF.
Product Description
Ergocalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ergocalciferol, 40,000,000 USP units/g
Supelco
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material