Merck
  • An automated device for screening the effects of psychotropic drugs on aggression and motor activity in mice.
An automated device for screening the effects of psychotropic drugs on aggression and motor activity in mice.
Pharmacology, biochemistry, and behavior (1980-08-01)
S Puglisi-Allegra, P Renzi
ABSTRACT
An automated technique for measuring simultaneously aggressive behavior and motor activity in mice is described. This technique allows measure of aggressive behavior and motor activity continuously throughout long-lasting periods without disturbing the animals except for feeding, watering and cleaning the cages. The effects of n-D-propylacetate, a drug which affects aggression without affecting motor activity, were also tested.
Sigma-Aldrich
Propyl acetate, ≥98%, FG
Sigma-Aldrich
Propyl acetate, natural, ≥97%, FCC, FG
Supelco
Propyl acetate, analytical standard
Sigma-Aldrich
Propyl acetate, 99%
Sigma-Aldrich
Propyl acetate, ≥99.5%