Merck
  • Why and how thioridazine in combination with antibiotics to which the infective strain is resistant will cure totally drug-resistant tuberculosis.
Why and how thioridazine in combination with antibiotics to which the infective strain is resistant will cure totally drug-resistant tuberculosis.
Expert review of anti-infective therapy (2012-10-04)
Leonard Amaral, Joseph Molnar
ABSTRACT
Over a period of 14 years, the authors have studied thioridazine, an old neuroleptic, that has been shown to have in vitro activity against intracellular Mycobacterium tuberculosis, regardless of its antibiotic resistance status, thioridazine cures infected mice of antibiotic-susceptible and multidrug-resistant tuberculosis (TB) infections and, when used in combination with antibiotics used for therapy of TB, renders the organism significantly more susceptible. This article will describe the authors' further work and the mechanisms by which thioridazine alone and in combination with antibiotics cures an extensively drug-resistant infection and why it is expected to cure totally drug-resistant TB infections as well. The concepts presented are entirely new and because they focus on the activation of killing by nonkilling macrophages where M. tuberculosis normally resides during infection, and coupled to the inhibition of efflux pumps which contribute to the antibiotic-resistant status, effective therapy of any antibiotic-resistant TB infection is possible.
Product Description
Thioridazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Supelco
Thioridazine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®