Merck
  • Evidence for mitoxantrone-induced block of inwardly rectifying K(+) channels expressed in the osteoclast precursor RAW 264.7 cells differentiated with lipopolysaccharide.
Evidence for mitoxantrone-induced block of inwardly rectifying K(+) channels expressed in the osteoclast precursor RAW 264.7 cells differentiated with lipopolysaccharide.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2012-08-03)
Chung-Lin Wang, Mei-Ling Tsai, Sheng-Nan Wu
ABSTRACT
Mitoxanthrone (MX) is an anthracenedione antineoplastic agent. Whether this drug and other related compounds have any effects on ion currents in osteoclasts remains largely unclear. In this study, the effects of MX and other related compounds on inwardly rectifying K(+) current (I(K(IR))) were investigated in RAW 264.7 osteoclast precursor cells treated with lipopolysaccharide. The I(K(IR))in these cells are blocked by BaCl(2) (1 mM). MX (1-100 µM) decreased the amplitude of I(K(IR)) in a concentration-dependent manner with an IC(50) value of 6.4 µM. MX also slowed the time course of I(K(IR)) inactivation elicited by large hyperpolarization. Doxorubicin (10 µM), 17β-estradiol (10 µM) and tertiapin (1 µM) decreased the I(K(IR)) amplitude in these cells. In bafilomycin A(1)-treated cells, MX-mediated block of I(K(IR)) still existed. In cell-attached configuration, when the electrode was filled with MX (10 µM), the activity of inwardly rectifying K(+) (Kir) channels was decreased with no change in single-channel conductance. MX-mediated reduction of channel activity is accompanied by a shortening of mean open time. Under current-clamp conditions, addition of MX resulted in membrane depolarization. Therefore, MX can interact with the Kir channels to decrease the I(K(IR)) amplitude and to depolarize the membrane in these cells. The block by this drug of Kir2.1 channels appears to be one of the important mechanisms underlying its actions on the resorptive activity of osteoclasts, if similar results occur in vivo. Targeting at Kir channels may be clinically useful as an adjunctive regimen to anti-cancer drugs (e.g., MX or doxorubicin) in influencing the resorptive activity of osteoclasts.
Sigma-Aldrich
Barium chloride dihydrate, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Barium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis