Merck
  • Attraction of Agrilus planipennis (Coleoptera: Buprestidae) to a volatile pheromone: effects of release rate, host volatile, and trap placement.
Attraction of Agrilus planipennis (Coleoptera: Buprestidae) to a volatile pheromone: effects of release rate, host volatile, and trap placement.
Environmental entomology (2012-06-27)
Krista L Ryall, Peter J Silk, Peter Mayo, Damon Crook, Ashot Khrimian, Allard A Cossé, Jon Sweeney, Taylor Scarr
ABSTRACT
Attraction of emerald ash borer, Agrilus planipennis Fairmaire, to a volatile pheromone was demonstrated in three field experiments using baited green sticky traps. A dose-response curve was generated for male A. planipennis to increasing release rates of (3Z)-dodecen-12-olide ((3Z)-lactone) in combination with the green leaf volatile, (3Z)-hexenol. Only the lowest release rate (<2.50 μg/d) of (3Z)-lactone significantly increased captures of male A. planipennis, as compared with traps baited with (3Z)-hexenol alone. Effect of trap height, (3Z)-lactone, and (3Z)-hexenol and their interactions on the trap capture of A. planipennis was determined in a factorial experiment. Number of males per trap was significantly and positively affected by (3Z)-lactone, (3Z)-hexenol, and trap height whereas number of females per trap was affected by trap height only; none of the interactions were significant. As predicted, the greatest mean catch of males was in traps baited with (3Z)-lactone and (3Z)-hexenol placed high in the canopy. Electroantennogram tests on the bark volatile, 7-epi-sesquithujene, demonstrated the ability of male and female A. planipennis antennae to detect and respond to this compound, particularly the (+)-7-epi-sesquithujene isomer. Results from an olfactometer bioassay and field testing did not demonstrate attraction of either males or females to (+)-7-epi-sesquithujene. These data increase our understanding of the pheromone ecology of the invasive emerald ash borer, provide further confirmation of the behavioral activity of the female-produced lactone pheromone, and should increase the ability to detect A. planipennis infestations where they are present.
Product Description
Supelco
cis-3-Hexen-1-ol, analytical standard
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%