Merck
  • Design and optimization of novel (2S,4S,5S)-5-amino-6-(2,2-dimethyl-5-oxo-4-phenylpiperazin-1-yl)-4-hydroxy-2-isopropylhexanamides as renin inhibitors.
Design and optimization of novel (2S,4S,5S)-5-amino-6-(2,2-dimethyl-5-oxo-4-phenylpiperazin-1-yl)-4-hydroxy-2-isopropylhexanamides as renin inhibitors.
Bioorganic & medicinal chemistry letters (2012-06-26)
Yuji Nakamura, Chie Sugita, Masaki Meguro, Shojiro Miyazaki, Kazuhiko Tamaki, Mizuki Takahashi, Yoko Nagai, Takahiro Nagayama, Mikio Kato, Hiroshi Suemune, Takahide Nishi
ABSTRACT
Introduction of the 2,2-dimethyl-4-phenylpiperazin-5-one scaffold into the P(3)-P(1) portion of the (2S,4S,5S)-5-amino-6-dialkylamino-4-hydroxy-2-isopropylhexanamide backbone dramatically increased the renin inhibitory activity without using the interaction to the S(3)(sp) pocket. Compound 31 exhibited >10,000-fold selectivity over other human proteases, and 18.5% oral bioavailability in monkey.
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Supelco
Piperazine hexahydrate, analytical standard