Merck
  • Differentiation of underivatized monosaccharides by atmospheric pressure chemical ionization quadrupole time-of-flight mass spectrometry.
Differentiation of underivatized monosaccharides by atmospheric pressure chemical ionization quadrupole time-of-flight mass spectrometry.
Rapid communications in mass spectrometry : RCM (2012-05-05)
Zhenqian Zhu, Liguo Song, John E Bartmess
ABSTRACT
Differentiation of underivatized monosaccharides is essential in the structural elucidation of oligosaccharides which are closely involved in many life processes. So far, such differentiation has been usually achieved by electrospray ionization mass spectrometry (ESI-MS). As an alternative to ESI-MS, atmospheric pressure chemical ionization mass spectrometry (APCI-MS) should provide complementary results. A quadrupole time-of-flight (QTOF) mass spectrometer with accurate mass measurement ability was used with an APCI heated nebulizer ion source because we believe that a recently published article using a single quadrupole mass spectrometer assigned incorrect identities for APCI ions from hexoses. Using APCI-QTOF, the MS(2) and pseudo-MS(3) mass spectra of 11 underivatized monosaccharides were obtained under various collision voltages. The mass spectra were carefully interpreted after accurate mass measurement. Differentiation of three hexoses was achieved by different MS(2) spectra of their [M + NH(4)](+) and [M - H](-) ions. The MS(2) spectra of the [M + NH(4)](+) ions were also used to distinguish methyl α-D-glucose and methyl β-D-glucose, while the pseudo-MS(3) spectra of the [M + H](+) ions were utilized to differentiate the three hexosamine and N-acetylhexosamine stereoisomers. Unique [M + O(2)](-) ions were observed and their distinctive fragmentation patterns were utilized to differentiate the three hexosamine stereoisomers. Although ESI coupled with single or triple quadrupole and ion trap mass spectrometers has been widely utilized in the differentiation of monosaccharides, this report demonstrated that APCI-QTOF-MS had its own advantages in achieving the same goal.
Product Description
Millipore
Methyl α-D-glucopyranoside, ≥99.0%, suitable for microbiology
Sigma-Aldrich
Methyl α-D-glucopyranoside, ≥99% (GC)
Sigma-Aldrich
Methyl β-D-glucopyranoside, ≥99% (HPLC and GC)