Merck
  • [Open population comparative randomized clinical trial of lavomax efficacy and safety in combined treatment of non-gonococcal urethritis].
[Open population comparative randomized clinical trial of lavomax efficacy and safety in combined treatment of non-gonococcal urethritis].
Urologiia (Moscow, Russia : 1999) (2011-01-25)
N V Kungurov, N M Gerasimova, N I Skidan, A P Gorbunov, N P Evstigneeva
ABSTRACT
A total of 40 patients with non-gonococcal urethritis (NGU) were divided into two groups. Twenty patients of group 1 received standard antibacterial treatment while 20 patients of group 2 received the same treatment plus an immunotropic drug based on tiloron (lavomax) in a course dose 1.25 g. Patients of group 2 had no recurrences while in group 1 recurrences were seen in 25% patients. Addition of lavomax resulted in clinical and etiological cure and activation of local (secretory IgA) and systemic (interferons alpha and gamma in blood serum) factors in anti-infection defense in NGU patients.
Sigma-Aldrich
Tilorone dihydrochloride, 95%