Merck
  • The separation of uranium ions by natural and modified diatomite from aqueous solution.
The separation of uranium ions by natural and modified diatomite from aqueous solution.
Journal of hazardous materials (2010-06-15)
Myroslav Sprynskyy, Iryna Kovalchuk, Bogusław Buszewski
ABSTRACT
In this work the natural and the surfactant modified diatomite has been tested for ability to remove uranium ions from aqueous solutions. Such controlling factors of the adsorption process as initial uranium concentration, pH, contact time and ionic strength have been investigated. Effect of ionic strength of solution has been examined using the solutions of NaCl, Na(2)CO(3) and K(2)SO(4). The pseudo-first order and the pseudo-second order models have been used to analyze the adsorption kinetic results, whereas the Langmuir and the Freundlich isotherms have been used to the equilibrium adsorption data. The effects of the adsorbent modification as well as uranium adsorption on the diatomite surface have been studied using X-ray powder diffraction, scanning electron microscopy and FTIR spectroscopy. The maximum adsorption capacities of the natural and the modified diatomite towards uranium were 25.63 micromol/g and 667.40 micromol/g, respectively. The desorptive solutions of HCl, NaOH, Na(2)CO(3), K(2)SO(4), CaCO(3), humic acid, cool and hot water have been tested to recover uranium from the adsorbent. The highest values of uranium desorption (86%) have been reached using 0.1M HCl.
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Filter agent, Celite® 545
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate