Merck
  • Kinetics for tautomerizations and dissociations of triglycine radical cations.
Kinetics for tautomerizations and dissociations of triglycine radical cations.
Journal of the American Society for Mass Spectrometry (2009-03-04)
Chi-Kit Siu, Junfang Zhao, Julia Laskin, Ivan K Chu, Alan C Hopkinson, K W Michael Siu
ABSTRACT
Fragmentations of tautomers of the alpha-centered radical triglycine radical cation, [GGG(*)](+), [GG(*)G](+), and [G(*)GG](+), are charge-driven, giving b-type ions; these are processes that are facilitated by a mobile proton, as in the fragmentation of protonated triglycine (Rodriquez, C. F. et al. J. Am. Chem. Soc. 2001, 123, 3006-3012). By contrast, radical centers are less mobile. Two mechanisms have been examined theoretically utilizing density functional theory and Rice-Ramsperger-Kassel-Marcus modeling: (1) a direct hydrogen-atom migration between two alpha-carbons, and (2) a two-step proton migration involving canonical [GGG](*+) as an intermediate. Predictions employing the latter mechanism are in good agreement with results of recent CID experiments (Chu, I. K. et al. J. Am. Chem. Soc. 2008, 130, 7862-7872).
Sigma-Aldrich
Gly-Gly-Gly
Sigma-Aldrich
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)