Merck
  • Boron trihalide mediated alkyne-aldehyde coupling reactions: a mechanistic investigation.
Boron trihalide mediated alkyne-aldehyde coupling reactions: a mechanistic investigation.
The Journal of organic chemistry (2008-03-06)
George W Kabalka, Min-Liang Yao, Scott Borella, Zhongzhi Wu, Yu-Hong Ju, Travis Quick
ABSTRACT
A boron trihalide mediated alkyne-aldehyde coupling reaction leading to stereodefined 1,3,5-triaryl-1,5-dihalo-1,4-pentadienes is described. The study led to the discovery of a direct substitution of hydroxyl groups by stereodefined alkenyl moieties using alkenylboron dihalides. During the investigation, it was also discovered that, at low temperatures, the reaction of BCl3 with alkynes produces monovinylboron dichloride rather than the reported divinylboron chloride. A modified reaction mechanism for the boron trichloride mediated alkyne-aldehyde coupling reaction is now proposed. The reaction temperature and mode of addition have been found to have dramatic affects on the stereochemistry of the diene products.
Sigma-Aldrich
Boron trichloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Boron trichloride solution, 1.0 M in p-xylene
Sigma-Aldrich
Boron trichloride solution, 1.0 M in heptane
Sigma-Aldrich
Boron trichloride solution, 1.0 M in toluene