Merck
  • Microwave-assisted synthesis of salicylamide via BCl3 mediated coupling.
Microwave-assisted synthesis of salicylamide via BCl3 mediated coupling.
Journal of combinatorial chemistry (2005-07-12)
Lei Zhang, John Y Zhang
ABSTRACT
A novel and efficient microwave-assisted, BCl3-mediated coupling reaction to synthesize salicylamide structures from phenols and isocyanates is described.
Sigma-Aldrich
Boron trichloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Boron trichloride solution, 1.0 M in p-xylene
Sigma-Aldrich
Boron trichloride solution, 1.0 M in heptane
Sigma-Aldrich
Boron trichloride solution, 1.0 M in toluene