Merck
First synthesis of etidronate partial amides starting from PCl3.
Organic & biomolecular chemistry (2003-10-07)
Petri A Turhanen, Riku Niemi, Mikael Peräkylä, Tomi Järvinen, Jouko J Vepsäläinen
ABSTRACT
Methods for the preparation of mixed tetra-amide esters 1 and 2, the partial amide ester 3, and tri- and P,P-diamides 4 and 5 from monophosphorus spieces 12, 8 and 9, respectively, were developed. Compounds 8 and 9 were obtained from phosphorus trichloride via MeOPCl2, which was treated with 2 eq. and 4 eq. of piperidine, followed by water or acetyl chloride, respectively. Tetrasubstituted amide bisphosphonates 1 and 2 were selectively dealkylated with lithium or silyl halide to achieve target compounds 3-5. Piperidine was found to be a good desilylation reagent. Quantum mechanical calculations illustrate why derivative 2 was produced in low yield. The usefulness of compounds 1, 3 and 4 as prodrugs of etidronate was determined in aqueous buffer and human serum.
Sigma-Aldrich
Phosphorus trichloride, 99.999% trace metals basis
Sigma-Aldrich
Phosphorus trichloride, ReagentPlus®, 99%