Merck
  • Pine weevil (Hylobius abietis) antifeedants from lodgepole pine (Pinus contorta).
Pine weevil (Hylobius abietis) antifeedants from lodgepole pine (Pinus contorta).
Journal of chemical ecology (2002-01-31)
K Bratt, K Sunnerheim, H Nordenhem, G Nordlander, B Langström
ABSTRACT
Pine weevils (Hylobius abietis) fed less on bark of lodgepole pine (Pinus contorta) than on bark of Scots pine (P. sylvestris). Two pine weevil antifeedants, ethyl trans-cinnamate and ethyl 2,3-dibromo-3-phenyl-propanoate, were isolated from bark of lodgepole pine. These two compounds significantly reduced pine weevil feeding in a laboratory bioassay. In field assays, the second compound significantly decreased pine weevil damage on planted seedlings. Ethyl 2,3-dibromo-3-phenylpropanoate has not previously been reported as a natural product.
Sigma-Aldrich
Ethyl cinnamate, natural, ≥95%, FG
Sigma-Aldrich
Ethyl cinnamate, 99%
Sigma-Aldrich
Ethyl cinnamate, ≥98%, stabilized, FCC, FG