Merck
  • Chlorine kinetic isotope effects on the haloalkane dehalogenase reaction.
Chlorine kinetic isotope effects on the haloalkane dehalogenase reaction.
Journal of the American Chemical Society (2001-07-18)
A Lewandowicz, J Rudziński, L Tronstad, M Widersten, P Ryberg, O Matsson, P Paneth
ABSTRACT
We have found chlorine kinetic isotope effects on the dehalogenation catalyzed by haloalkane dehalogenase from Xanthobacter autotrophicus GJ10 to be 1.0045 +/- 0.0004 for 1,2-dichloroethane and 1.0066 +/- 0.0004 for 1-chlorobutane. The latter isotope effect approaches the intrinsic chlorine kinetic isotope effect for the dehalogenation step. The intrinsic isotope effect has been modeled using semiempirical and DFT theory levels using the ONIOM QM/QM scheme. Our results indicate that the dehalogenation step is reversible; the overall irreversibility of the enzyme-catalyzed reaction is brought about by a step following the dehalogenation.
Sigma-Aldrich
1-Chlorobutane, biotech. grade, for protein sequence analysis, ≥99.8%
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
1-Chlorobutane, suitable for HPLC, ≥99.8%
Sigma-Aldrich
1-Chlorobutane, anhydrous, 99.5%